Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Acurex™ 501 and 502 Compact Bottle-Top Dispensers, DWK Life Sciences
Supplier: DWK Life Sciences
Acurex™
W844055 844064 W844057 844068 844060 W844058 W844056 W844059
53501-632EA 1035.06 USD InStock
53501-632 89044-740 53501-634 14231-604 89044-736 89044-738 89044-732 89044-734
Acurex™ 501 and 502 Compact Bottle-Top Dispensers, DWK Life Sciences
Bottle Top Dispensers
Compact dispensers for safe reagent handling, adapted to refrigerator storage as well as water bath heating
Dosing mechanism entirely protected inside reservoir. Manufactured and tested to fully comply with the latest regulations on instrument safety and precision. Volumes range from 0.2 to 30 mL. This product is also fully autoclavable at 121°C (250°F).
Ordering information: One-year manufacturer's warranty.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty