Would you like to log in again?
SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
SOCOREX® LAB 45™
W832022 W832018 W832024
89044-836EA 27.61 USD InStock
89044-836 89044-838 89044-832
SOCOREX® LAB 45™ Reagent Bottles, DWK Life Sciences
Bottles Plastic Bottles
Natural HDPE bottles with screw cap.
Perfectly adapted with Calibrex™ bottle top dispensers. Corresponding neck adapters supplied with the dispensers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty