Would you like to log in again?
Boston Round Bottles, Amber, Narrow Mouth, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5121628C-26 5120824V-21 5120120C-24 5121628V-25 5120120V-21 5120220B 5120220C-26 5120422V-26 5120824V-25 5120422B 5121628B 5120824B 5120120V-26 5120422C-26 5120422V-21 5120120B 5121628C-24 5120824C-26 5120220C-24 5121628V-22 5121628V-21 5120824C-24 5120824C-25 5120220V-25 5123233C-25 5121628C-25 5123233V-22 5123233V-21 5120220V-21 5123233V-26 5120120V-25 5121628V-26 5120220V-26 5120120C-26 5120422V-25 5120220C-25 5120120C-25 5120824V-26 5120422C-24 5123233C-24 5123233V-25 5120422C-25 5123233B 5123233C-26
89045-392CS 145.52 USD InStock
89045-392 89045-370 89045-390 89045-274 89045-396 89045-374 89045-272 89045-394 89045-372 89045-278 89045-410 89045-356 89045-378 89045-276 89045-376 89045-398 89045-414 89045-412 89045-358 89045-418 89045-416 89045-380 89045-282 89045-280 89045-362 89045-384 89045-382 89045-360 89045-388 89045-366 89045-422 89045-400 89045-364 89045-386 89045-420 89045-304 89045-404 89045-426 89045-368 89045-424 89045-402 89045-408 89045-306 89045-406
Boston Round Bottles, Amber, Narrow Mouth, Kimble®, DWK Life Sciences
Bottles Glass Bottles
High quality, economical narrow mouth bottles are manufactured from Type III soda-lime glass.
The amber tinting protects light-sensitive contents. Supplied with black phenolic caps.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are also packaged in bulk packs. Pallet packs consist of shrink-wrapped glassware bundles stacked onto shrink-wrapped pallets. This packaging is designed for large glassware quantities.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty