Would you like to log in again?
Straight-Sided Jars, Clear Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
5410870C-23 5410663V-26 5413289C-22 5410663C-23 5410663V-22 5410458V-21 5413289C-21 5410253V-21 5410870V-26 5413289B 5410870B 5410458V-22 5413289V-21 5410663C-21 5410253V-22 5410253V-24 5410253V-26 5411689C-24 5410458C-26 5410253C-26 5411689C-21 5411689C-26 5410253C-21 5410458B 5410458C-21 5410663C-26 5410870C-24 5411689V-22 5410253B 5411689B 5410253C-23 5411689V-21 5410458V-26 5413289V-22 5411689V-26 5410870C-26 5410663B 5410870V-21 5413289V-26 5410870C-21 5410253C-24 5410663V-21 5410458C-24 5413289C-24 5410870V-22 5410870C-22 5410663C-24 5410458C-23 5411689C-22 5410458V-24 5413289C-26
89045-770CS 129.63 USD InStock
89045-770 89045-850 89045-774 89045-772 89045-854 89045-778 89045-852 89045-776 89045-858 89045-814 89045-812 89045-856 89045-818 89045-816 89045-780 89045-784 89045-862 89045-782 89045-860 89045-866 89045-786 89045-864 89045-826 89045-824 89045-868 89045-872 89045-752 89045-796 89045-870 89045-794 89045-876 89045-756 89045-874 89045-798 89045-754 89045-836 89045-878 89045-758 89045-838 89045-880 89045-762 89045-840 89045-760 89045-882 89045-766 89045-844 89045-764 89045-842 89045-848 89045-768 89045-846
Straight-Sided Jars, Clear Glass, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Jars feature a wide-mouth design for efficient addition and removal contents
Clear glass allows for easy viewing of contents. Made of Type III soda-lime glass. Available with or without black phenolic closures.
Ordering information: Packaging options include bulk packs (shrink-wrapped modules with caps in bags), convenience packs (caps attached), or pallet packs (jars only; caps not included). All 1L (33.8oz.) jars are packed in corrugated cartons with divider cells. Pallet packs of 1L (33.8oz.) jars contain 54 cartons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty