Would you like to log in again?
Straight-Sided Jars, Amber Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
5420458V-21 5420870C-26 5420870V-21 5421689C-26 5420458C-26 5420458V-26
89046-010CS 109.03 USD InStock
89046-010 89046-012 89045-994 89045-898 89046-006 89045-902
Straight-Sided Jars, Amber Glass, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Jars feature a wide-mouth design for efficient addition and removal contents
Amber-colored glass protects light-sensitive contents. Made of Type III soda-lime glass. Available with or without black phenolic closures.
Ordering information: Packaging options include bulk packs (shrink-wrapped modules with caps in bags), convenience packs (caps attached), or pallet packs (jars only; caps not included). All 1L (33.8oz.) jars are packed in corrugated cartons with divider cells. Pallet packs of 1L (33.8oz.) jars contain 54 cartons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty