Would you like to log in again?
Straight-Sided Tall Jars, Clear Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
5513289C-81 5510858V-86 5513289B 5511670V-86 5510858B 5510448V-86 5510448B 5511670V-81 5513289C-86 5513289C-82 5510448V-81 5511670C-81 5513289V-81 5511670B 5510858V-81 5510448C-81 5510858C-81 5513289V-86
89046-040CS 179.42 USD InStock
89046-040 89046-052 89046-042 89046-054 89046-120 89046-044 89046-110 89046-122 89046-046 89046-124 89046-048 89046-126 89046-104 89046-106 89046-118 89046-108 89046-070 89046-050
Straight-Sided Tall Jars, Clear Glass, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Jars feature a wide-mouth design for efficient addition and removal contents
Clear glass allows for easy viewing of contents. Made of Type III soda-lime glass. Available with or without black phenolic closures.
Ordering information: Packaging options include bulk packs (shrink-wrapped modules with caps in bags), convenience packs (caps attached), or pallet packs (jars only; caps not included). All 1L (33.8oz.) jars are packed in corrugated cartons with divider cells. Pallet packs of 1L (33.8oz.) jars contain 54 cartons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty