Would you like to log in again?
French Square Bottles, Clear, Wide Mouth, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMBLE®
5610124C-24 5610433V-21 5610228V-25 5610228C-24 5610124C-25 5610124V-21 5611648V-22 5610228C-22 5611648C-22 5610433B 5610843V-21 5610843B 5610228C-21 5611220C-25 5610228V-26 5611220C-22 5610124V-25 5610433V-25 5611220V-21 5610433C-24 5611220V-26 5611648B 5611648C-24 5610124B 5610433V-26 5610433C-21 5611648C-21 5610843C-21 5610228C-25 5611648V-26 5611220V-25 5611220C-24 5610433C-22 5610124C-21 5610228V-21 5611220B 5611648V-21 5611648V-24 5610843C-24 5610843V-26 5610228B 5610843C-22 5610124C-22 5610124V-26 5611220C-21 5610433C-25 5610843V-22
89046-172CS 1018.35 USD InStock
89046-172 89046-150 89046-272 89046-250 89046-152 89046-174 89046-252 89046-230 89046-274 89046-176 89046-154 89046-254 89046-232 89046-276 89046-156 89046-178 89046-278 89046-234 89046-256 89046-136 89046-158 89046-236 89046-258 89046-138 89046-238 89046-218 89046-170 89046-282 89046-140 89046-240 89046-142 89046-242 89046-220 89046-144 89046-222 89046-244 89046-146 89046-168 89046-224 89046-246 89046-148 89046-226 89046-248 89046-228 89046-180 89046-280 89046-182
French Square Bottles, Clear, Wide Mouth, KIMBLE®, DWK Life Sciences
Bottles Glass Bottles
Clear, sturdy bottles are constructed of Type III soda-lime glass and feature a wide mouth design to facilitate the addition and removal of sample contents.
  • Unless otherwise noted, bottles are supplied with black phenolic caps
The clear glass design allows for constant viewing of contents. Square bottle shapes help to maximize storage and shelf space.
Packaging: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are also packaged in bulk packs.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty