Would you like to log in again?
Standard Jars, Clear, Wide Mouth, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
5310858V-21 5310448C-26 5310448V-21 5310448V-26 5310858C-26 5310858V-26
89046-296CS 150.36 USD InStock
89046-296 89046-330 89046-298 89046-332 89046-334 89046-328
Standard Jars, Clear, Wide Mouth, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Standard jars are manufactured from Type III soda-lime glass
The clear glass design features rounded shoulders and permits easy viewing of contents. Wide-mouth openings allow for easy addition and removal of contents. Jars are supplied with screw-on caps unless otherwise noted.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty