Would you like to log in again?
Glass Jugs, Amber and Clear, KIMBLE® DWK Life Sciences
Supplier: DWK Life Sciences
5919138V-26 5916438V-25 5928038B 5928038V-26 5916438B 5919138B 5928038V-25 5916438V-26 5919138V-25
89046-938CS 283.09 USD InStock
89046-938 89046-918 89046-930 89046-920 89046-942 89046-910 89046-922 89046-912 89046-924
Glass Jugs, Amber and Clear, KIMBLE® DWK Life Sciences
Bottles Carboys
Jugs are made of Type III soda-lime glass and feature narrow mouths and sturdy handles for safe pouring.
  • With or without black phenolic closures
Amber glass protects light-sensitive contents. Clear glass provides easy viewing of contents.
Ordering information: All jugs are packaged in corrugated cartons with divider cells and caps attached.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty