Would you like to log in again?
Square Tablet Bottles, Glass, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences
5910133C-25 5910133C-26 5910133B 5910133C-21
89046-958CS 890.62 USD InStock
89046-958 89046-950 89046-954 89046-946
Square Tablet Bottles, Glass, KIMBLE®, DWK Life Sciences
Bottles Glass Bottles
Square tablet bottles facilitate small sample collection and storage.
Constructed of clear Type III soda-lime glass, bottles allow for constant sample viewing. The wide mouth design aids in efficient addition and removal of contents. An assortment of cap and liner options are available. All bottles include caps unless otherwise noted.
Ordering information: Bottles are packaged in corrugated cartons with divider cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty