Would you like to log in again?
Testing Jars, Clear, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
5910243C-26 5910243C-21 5910243B
89046-962CS 661.34 USD InStock
89046-962 89046-964 89046-966
Testing Jars, Clear, Kimble Chase, DWK Life Sciences
Jars General Use Jars
Jars feature a wide-mouth design for efficient addition and removal contents
Clear glass allows for easy viewing of contents. Made of Type III soda-lime glass. Packaged in bulk packs (shrink modules with caps in bags).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty