Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sleeve-Style Rubber Stoppers, Wheaton
Supplier: DWK Life Sciences
224100-320 224100-160 224100-060 224100-020 224100-161
89047-006CS 858.42 USD InStock
89047-006 16170-462 89047-004 80093-832 80093-834
Sleeve-Style Rubber Stoppers, Wheaton
Stoppers Rubber Stoppers
Plug stoppers with fold-over skirts are for use on serum vials and other crimp-finish containers that do not require an aluminum seal or overcap
Stoppers may also be used as injection sites for multiple dose entry. General-purpose natural red rubber is excellent for resealing and multi-puncture applications. Diameter: 100mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty