Would you like to log in again?
VWR® Signature™ Disposable Electroporation Cuvettes
Supplier: VWR
VWR® Signature™
4040-04 1040-04S 4010-01 1010-01 1040-04 4020-02 1020-02
89047-208CS 11412.03 USD InStock
89047-208 89047-206 76102-580 76102-576 89047-210 76407-204 76102-578
VWR® Signature™ Disposable Electroporation Cuvettes
Cuvettes Electroporation Cuvettes
VWR Signature™ Disposable Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells.
  • Cuvettes are precision built to provide exacting tolerances, ensuring reproducible results
  • Polished aluminum plates reduce static and arcing
  • Compatible with most electroporation devices
  • Gamma irradiated for sterility
Disposable cuvettes are manufactured from high quality, durable polycarbonate material that has been tested to optimize transformation efficiencies.
Ordering information: Each pack contains 50 individually wrapped cuvettes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty