Would you like to log in again?
Rtx®-Wax GC Columns, Restek
Supplier: Restek
Rtx®
12458 12423 12436 12435 12426 12424 12408 41602 12455 12451 12438 12452 41603 12455 12441 12457 12440 12420 12439 41604 12442 12421 12454 12427
10059-538EA 744.79 USD InStock
10059-538 89048-608 AP400000-002 89048-626 89048-616 89048-638 89048-606 89048-628 89048-618 89048-644 89048-622 89048-600 89048-612 89048-634 89048-624 89048-602 89048-636 89048-614 89048-596 89048-640 89048-630 89048-642 10059-542 10059-540
Rtx®-Wax GC Columns, Restek
Chromatography Columns
Highly efficient and inert general-purpose PEG columns are ideal for FAMEs, solvents, BTEX aromatics, and flavor compounds.
  • Excellent selectivity for compounds of intermediate to high polarity
  • Equivalent to USP G14, G15, G16, G20, and G39 phases
  • Temperature range: 20 to 250 °C (68 to 482 °F)
Columns feature an extended operating temperature range to allow for the analysis of compounds having a wide volatility range, and to ensure low bleed at temperatures as high as 250 °C (482 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty