Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CELLMASTER™ Cell Culture Roller Bottles, PET, Standard Screw Cap, Sterile, Greiner Bio-One

Supplier: Greiner bio-one
CELLMASTER™
680180 680170
89047-262CS 168.75 USD InStock
89047-262 89049-110
CELLMASTER™ Cell Culture Roller Bottles, PET, Standard Screw Cap, Sterile, Greiner Bio-One
Bottles Roller Bottles
Cell culture roller bottles are available in short with a smooth or a ribbed surface.

  • Surface suitable for many adherent cell lines
  • PET is very stable and gas permeable
  • Break-proof
  • All bottles have standard screw caps

The ribbed design increases the growth area of the roller bottle without changing the height.

Sterilized by irradiation (SAL 10⁻³). USP Class VI certified. DNase-, RNase-, and human DNA-free, non-cytotoxic (EN ISO 10993) and non-pyrogenic.

Short form bottles with a ribbed design have a 2-fold or 2.5-fold expanded growth area. All bottles are certified free of RNase, DNase and human DNA, are nonpyrogenic and noncytotoxic.

Bottles are clear with blue cap. Bottles with a 100 to 1000 ml range have white graduation print and writing area. The bottle with 100 to 2000 ml range has black graduation print and writing area.

Temperature range:
Bottle: –60 to 150°C (–76 to 302°F)
Cap: –50 to 100°C (5 to 212°F)

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty