Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quartz Crucibles and Lids, Technical Glass Products

Supplier: Technical Glass Products (TGP)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FQVWR10CRU FQVWR15LID FQVWR150LID FQVWR50CRU FQVWR20CRU FQVWR30LID FQVWR30CRU FQVWR15CRU FQVWR100CRU FQVWR10LID FQVWR20LID FQVWR150CRU FQVWR50LID FQVWR100LID
89049-630EA 125.82 USD InStock
89049-630 89049-620 89049-612 89049-624 89049-632 89049-610 89049-622 89049-616 89049-606 89049-628 89049-614 89049-626 89049-618 89049-608
Quartz Crucibles and Lids, Technical Glass Products
Crucibles
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%

Quartz crucibles and lids are invaluable in research and industry. Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These properties include; extremely high purity, high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.

Ordering information: All lids are sold separately.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty