Would you like to log in again?
Microscope Slides, Frosted Glass, Azer Scientific
Supplier: Azer Scientific
VWR®
EMS200WS EMS200BS
89049-672CS 431.16 USD InStock
89049-672 89049-670
Microscope Slides, Frosted Glass, Azer Scientific
Slides Microscope Slides
VWR frosted microscope slides are specifically designed for compatibility with Sysmex® systems, they are manufactured from high-quality glass. These slides are pre-cleaned with smooth grounded edges and undergo continuous premium quality processing. All slides have single side, color frosted ends which helps to avoid sticking and jamming problems that can occur with other slides. The color frosted end is resistant to all common chemicals that are utilized in the laboratory. Slides feature 45° clipped corners for ease in handling, and two choices of color-coded frosted ends to aid in quick and simple identification.
  • Clipped corners at 45°
  • Single-frosted edges
  • Sysmex® systems compatible
  • High-quality glass
They are manufactured from high-quality glass. Slides are pre-cleaned with smooth grounded edges and undergo continuous premium quality processing. All slides have single side, color frosted ends which help to avoid sticking and jamming problems that can occur with other slides. The color frosted end is resistant to all common chemicals that are utilized in the laboratory. Slides feature 45° clipped corners for ease in handling, and two choices of color-coded frosted ends to aid in quick and simple identification.
Packaging: 72 per box, 20 boxes per case
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty