Would you like to log in again?
ThermoSafe® Absorbent Sheets, Sonoco ThermoSafe
Supplier: Sonoco
ThermoSafe®
ZORB1212 ZORB66 ZORB33
89050-004CS 589.43 USD InStock
89050-004 89050-006 89050-008
ThermoSafe® Absorbent Sheets, Sonoco ThermoSafe
Spill Control Materials
Sheets absorb spills that may occur during shipment of diagnostic specimens and biological materials.
  • Spill control material
  • Meets packaging and transportation regulations
Made of tissue-based paper substrate impregnated with super-absorbent polymers to help prevent leakage and possible contamination of package handler. Sheets meet regulations and recommendations of U.S. Government, U.S. Postal Service, and United Nations for goods packaging and transporation of diagnostic specimens, biological materials, and other dangerous goods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty