Would you like to log in again?
Superfrost® Cytogenetics Slides, Erie Scientific (Thermo Scientific)
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Superfrost®
4951-7140-006
89050-838CS 913.48 USD InStock
89050-838
Superfrost® Cytogenetics Slides, Erie Scientific (Thermo Scientific)
Slides Microscope Slides
The consisetency and quality of these slides allows for reliable chromosome spreads, while the hydrophilic surface improves retention of metaphase cells
Slides have a Superfrost® white tab for permanent marking.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty