Would you like to log in again?
Adapter, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5155-10
89051-296EA 645 USD InStock
89051-296
Adapter, Jacketed, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is jacketed with water-cooled 24/40 standard taper joints.
  • Extension arm below joint
  • 105° angle
  • Jacketed
  • Fabricated from borosilicate glass
The extension arm below the joint is 50mm. Use with a 3/8-inch I.D. tubing, size D hose connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty