Would you like to log in again?
Adapter, 75 Degree Angle, Ace Glass Incorporated
Supplier: Ace Glass
5092-54
89051-316EA 128.16 USD InStock
89051-316
Adapter, 75 Degree Angle, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a standard taper inner joint at the bottom and side, with a #7 Ace-Thred joint at the top.
  • Bottom and side [ST] inner joints
  • Top Ace-Thred
  • Nylon bushing and FETFE o-ring included
  • Fabricated from borosilicate glass
Adapter comes with a 24/40 standard taper inner joint at the bottom and side. The top joint has a #7 Ace-Thred. Supplied complete with 89051-122 nylon bushing and an 89060-958 FETFE o-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty