Would you like to log in again?
Glass Pouring Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5120-14 5120-20
89427-892EA 62.11 USD InStock
89427-892 89051-332
Glass Pouring Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This convenient adapter is designed for pouring from flasks or funnels without pouring over a ground joint.
  • Conveniently pour from flask or funnel
  • Bottom [ST] inner joint
  • Fabricated from borosilicate glass
Each adapter features a 24/40 standard taper inner joint at the bottom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty