Would you like to log in again?
Adatper, Vacuum Take-Off, Long Stem, Ace Glass Incorporated
Supplier: Ace Glass
9124-06 5195-15 5195-10
89051-380EA 136 USD InStock
89051-380 89051-378 89074-286
Adatper, Vacuum Take-Off, Long Stem, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is designed for use with distillation set-ups.
  • 105° angle outer joint
  • Long stem
  • With hose connection
  • Fabricated from borosilicate glass
Featuring an outer joint that is at an angle of 105°. The hose connection is on the side opposite of the upper joint. Length values represent the stem length below bottom joint (mm).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty