Would you like to log in again?
Hose Connection Adapter, [SJ], Ace Glass Incorporated
Supplier: Ace Glass
5216-23 5216-35
89051-426EA 59.49 USD InStock
89051-426 89051-424
Hose Connection Adapter, [SJ], Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These adapters feature a straight hose connection at the top.
  • Bottom spherical ball joint
  • Top hose connection
  • Fabricated from borosilicate glass
Adapters have a spherical ball joint at the bottom and a straight hose connection at the top.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty