Would you like to log in again?
Adapter, Firestone, with Fritted Disc, Ace Glass Incorporated
Supplier: Ace Glass
5257-53 5257-62 5257-43 5257-49
89051-462EA 183.91 USD InStock
89051-462 89051-460 89051-466 89051-464
Adapter, Firestone, with Fritted Disc, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This rotary evaporator “splash guard” adapter features a coarse Porosity A fritted disc that assures there will be no carry-over in the event of splash-up.
  • Top and bottom [ST] medium length joints
  • Coarse Porosity A fritted disc
  • Fabricated from borosilicate glass
The adapter's standard taper medium length joints, top and bottom, will accept full length standard taper joints. The coarse Porosity A (147-174 micron) fritted disc assures there will be no carry-over in the event of splash-up. Overall length is kept to a minimum to effect best distillation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty