Would you like to log in again?
Mini #7 Ace-Thred with Hose Connection Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5261-16 5261-08 5261-12 5261-06 5261-20
89064-828EA 83.44 USD InStock
89064-828 89064-826 89064-824 89051-406 89051-498
Mini #7 Ace-Thred with Hose Connection Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "Mini" adapter has a standard taper inner joint at the bottom, #7 Ace-Thred at the top and a serrated hose connection.
  • Ace-Thred at top
  • Bottom [ST] inner joint
  • Serrated hose connection
  • Fabricated from borosilicate glass
Adapter will accommodate thermometers, and bleed tubes up to 7mm in diameter. Suitable for most vacuum work.
Ordering information: Supplied complete with 89051-122 nylon bushing and 89060-958 FETFE o-ring.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty