Would you like to log in again?
Adapter, [SJ], Straight Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5300-26
89051-564EA 150.8 USD InStock
89051-564
Adapter, [SJ], Straight Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a straight hose connection with a spherical ball joint at the bottom and glass stopcock.
  • Straight hose connection
  • Bottom spherical ball joint
  • Glass stopcock
  • Fabricated from borosilicate glass
Adapter has a glass stopcock with a 2mm plug bore. Bore size represents the plug bore size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty