Would you like to log in again?
Bottle, Specific Gravity, Ace Glass Incorporated
Supplier: Ace Glass
5415-05 5415-06 5415-16 5415-12 5415-08
89051-680EA 206.63 USD InStock
89051-680 89078-380 89051-688 89051-686 89051-684
Bottle, Specific Gravity, Ace Glass Incorporated
Specific Gravity Bottles
This conical shape flask is designed for maximum stability.
Complete item is supplied with non-mercury thermometers for accurate reading, and standard taper 5/12 cap for side arm. Thermometer has a range of 14 to 38ºC, 0.2ºC subdivisions, with standard taper 10/18 joint at bottom. Thermometer is a 25mm immersion. Fabricated from borosilicate glass.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty