Acrylic safety shield and polyethylene cap for use with Michel-Miller columns to insure safe operation at pressures to 300 psi
Top thread of column is hung in polyethylene cap; cap is then held by the plastic shield. Clamp the shield, not the column. Wall thickness of shield is 3 mm.