Would you like to log in again?
Stopper, Michel-Miller, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5803-25 5803-11 5803-05 5803-08
89052-110EA 118.09 USD InStock
89052-110 89052-108 89052-112 89052-114
Stopper, Michel-Miller, Ace Glass Incorporated
Stoppers PE/LDPE/HDPE Stoppers
These precision made PTFE stoppers are for use at either end of Michel-Miller columns, or on other Ace-Thred connections, when a leak-tight seal without the use of o-rings is desired
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty