Would you like to log in again?
Adapter, Michel-Miller, Injection Port, Ace Glass Incorporated
Supplier: Ace Glass
5807-49 5807-14 5807-48 5807-18
89052-140EA 191.79 USD InStock
89052-140 89052-142 89052-138 89052-144
Adapter, Michel-Miller, Injection Port, Ace Glass Incorporated
Fittings and Connectors
A simple, all-PTFE septum injector that allows for the most efficient use of Michel-Miller non-packed columns.
  • Direct on-column injection
  • Leak-tight seal without o-rings
  • Threaded
Injector allows direct on-column injection to the center of the filter disc at top of column without stopping the solvent flow. Designed to make a leak-tight seal without the use of o-rings. Simply tighten until white ring appears indicating a seal has been made, then secure locknut; no need to over tighten. Top thread is 5/16-inch-24 UNF for use with injection port cap that holds a septum. Male thread is size designation and matches thread size of columns. Bore is 1.5mm (.060-inches).
Ordering information: Adapter is available with or without Heli-Coil insert. Supplied with one PTFE coated silicone rubber septum. Use paper filter discs (not included) to retain packing material. Not suitable for larger chromatography columns. Contact your Avantor representative for details.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty