Would you like to log in again?
Adapter, Column, Feed Tube, Ace Glass Incorporated
Supplier: Ace Glass
5826-30 5826-28 5826-20 5826-24
89052-456EA 127.12 USD InStock
89052-456 89052-458 89052-452 89052-454
Adapter, Column, Feed Tube, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is threaded at both ends.
  • Top is #7 Ace-Thred
  • Lower thread
  • Fabricated from borosilicate glass
The top of this adapter has a #7 Ace-Thred that is supplied with a nylon bushing and FETFE o-ring for use with adjustable feed tubes. Size listed also refers to the I.D. of the tubing above the lower thread, and this corresponds to the column diameter.
Ordering information: For extra bushings (includes two FETFE o-rings), order 89051-122. For o-ring only, order 89060-958 (package of 12).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty