Would you like to log in again?
Adapter, Bottom Drip, with NPT Thread, Ace Glass Incorporated
Supplier: Ace Glass
5838-75 5838-80 5838-82 5838-83 5838-72 5838-76 5838-78
89052-592EA 120.55 USD InStock
89052-592 89052-594 89052-590 89052-600 89052-588 89052-596 89052-598
Adapter, Bottom Drip, with NPT Thread, Ace Glass Incorporated
Fittings and Connectors
For use as a column end fitting that allows connection of appropriate size Ace-Thred to tubing connectors with NPT threads.
  • NPT thread
  • Fabricated of PTFE
  • Supplied complete with disc support, screen support with retainer ring, and o-ring
Adapters are fabricated of PTFE and supplied complete with one 100 micron polyethylene disc support, one 350 micron polypropylene screen support with PTFE retainer ring, and FETFE or CAPFE o-ring. The screen with retainer ring support has a large pore size and offers less resistance to flow. However, when smaller particle size packing is used, it will be necessary to replace the screen and retainer ring with the 100 micron support.
Ordering information: Glass filter discs are also available. Contact your Avantor representative for further information. For optional filter disc remover, order 89052-532.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty