Would you like to log in again?
Adapter, Bottom Drip, NPT Thread, with Glass Disc, Ace Glass Incorporated
Supplier: Ace Glass
5838-91 5838-94 5838-96
89052-604EA 229.7 USD InStock
89052-604 89052-606 89052-602
Adapter, Bottom Drip, NPT Thread, with Glass Disc, Ace Glass Incorporated
Fittings and Connectors
For use as a column end fitting that allows connection of appropriate size Ace-Thred to tubing connectors with NPT threads.
  • Glass disc
  • Fabricated of PTFE
  • Supplied complete with one glass filter disc and one CAPFE o-ring
Adapters are fabricated of PTFE and supplied complete with one 25-50 micro Porosity C glass filter disc and one CAPFE o-ring.
Ordering information: For optional filter disc remover, order 89052-532.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty