Would you like to log in again?
Ace-Thred Coupling, Nylon or PTFE, Back Seal, Ace Glass Incorporated
Supplier: Ace Glass
5840-15 5840-05 5840-49 5840-20 5840-10 5840-60 5840-47 5840-51 5840-45
89052-626EA 103.13 USD InStock
89052-626 89052-648 89052-636 89052-638 89052-632 89052-536 89052-624 89052-562 89052-652
Ace-Thred Coupling, Nylon or PTFE, Back Seal, Ace Glass Incorporated
Fittings and Connectors
Used to couple threading columns together, or to end fittings with single o-ring seal for leak-tight engagement and with no significant size reduction in I.D
This coupling has "back seals", which means that the o-ring on the Ace-Thred connections is located at the TOP of the threads rather than at the bottom. For a better, more consistent seal, consider using "front seals" instead. Front seals are recommended for critical pressure applications, and they also prevent viscous materials from migrating up into the threads.
Can also be used for coupling manifolds. Ace-Thred size listed refers to inside diameter of threads and corresponds to the column diameter.
Ordering information: Supplied with two FETFE o-rings. FETFE is an Ace Glass exclusive compound, a fluoroelastomer with TFE additives. It has good compression set along with excellent temperature and chemical compatibility. FETFE is a USP Class VI material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty