Would you like to log in again?
Ace-Thred Reducing Coupling, Ace Glass Incorporated
Supplier: Ace Glass
5842-48 5842-46 5842-05 5842-15 5842-10 5842-50
89052-680EA 151.34 USD InStock
89052-680 89052-660 89052-670 89052-666 89052-656 89052-662
Ace-Thred Reducing Coupling, Ace Glass Incorporated
Fittings and Connectors
These couplings are designed for threading columns together, or to secure end fittings with a single O-Ring seal for leak-tight engagement, with no significant size reduction in the I.D
One end is threaded for the next smallest diameter. Size listed refers to inside diameter of threads and corresponds to the column diameter. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty