Would you like to log in again?
Plug Stoppers with Back Seal, Nylon or PTFE, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5845-43 5845-51 5845-45 5845-49 5845-47 5845-243 5845-12 5845-17 5845-05 5845-50 5845-15 5845-30 5845-10 5845-56 5845-03 5845-48 5845-20
89052-816EA 80.96 USD InStock
89052-816 89052-808 89052-818 89084-368 89052-856 89052-834 89052-812 89084-366 89052-814 89052-836 89052-858 89052-852 89052-800 89052-854 89052-832 89052-810 76318-504
Plug Stoppers with Back Seal, Nylon or PTFE, Ace Glass Incorporated
Stoppers PE/LDPE/HDPE Stoppers
Solid plug stoppers are designed for sealing column ends.
Plug stoppers feature a "back seal," meaning the O-Ring on the Ace-Thred connection is located at the top of the threads.
They allow for the preparation and storage of columns. Each stopper is supplied with either one FETFE O-Ring or one Silicone O-Ring. Available in Nylon or PTFE construction, with the exception of item 89052-812, which is fabricated from Ultra High Molecular Weight Polyethylene (UHMWPE).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty