Would you like to log in again?
Plug Stoppers with Front Seal, Nylon or PTFE, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5846-51 5846-44 5846-50 5846-04 5846-14 5846-60 5846-45 5846-49 5846-06 5846-22 5846-27 5846-46 5846-12 5846-16 5846-52 5846-48 5846-18
89084-370EA 52.59 USD InStock
89084-370 89052-828 89052-806 89052-838 89052-846 89052-802 89052-824 89052-804 89052-826 89052-848 89052-820 89052-842 89052-830 89052-822 89052-844 89052-840 89052-850
Plug Stoppers with Front Seal, Nylon or PTFE, Ace Glass Incorporated
Stoppers PE/LDPE/HDPE Stoppers
Solid plug stoppers are designed for sealing column ends
Plug stoppers feature a "front seal," meaning the O-Ring on the Ace-Thred connection is located at the bottom of the threads rather than the top. This provides a better, more consistent seal. This is critical for pressure applications, and it also prevents viscous materials from migrating up into the threads.
They permit preparation and storage of columns. Each stopper is supplied with one FETFE O-Ring, with the exception of item 89084-370, which is supplied with a CAPFE O-Ring. Plug stoppers are available in Nylon or PTFE, with the exception of item 89052-842, which is fabricated from Ultra High Molecular Weight Polyethylene (UHMWPE).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty