Would you like to log in again?
Needles, Chromatography, Syringe, Ace Glass
Supplier: Ace Glass
5936-44 5936-39 5936-32 5936-40
89053-772PK 130.49 USD InStock
89053-772 89053-770 89053-776 89053-774
Needles, Chromatography, Syringe, Ace Glass
Syringe Needles
These standard hypodermic type needles are made from 304 full hard stainless steel tubing with chrome plated brass American standard Luer-Lok taper short hubs
Supplied 50 mm (2") long with point style #2 (20° bevel) for septum penetration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty