Would you like to log in again?
Condenser, Friedrichs, 200 mm, Ace Glass Incorporated
Supplier: Ace Glass
5969-10
89053-992EA 671.25 USD InStock
89053-992
Condenser, Friedrichs, 200 mm, Ace Glass Incorporated
Condensers
This Friedrichs condenser is reflux controlled by regulating the jacket temperature
Provides a compact, spiral path for condensation. Bottom is a standard taper 24/40 inner joint. Jacket length is 200 mm. Use with 3/8-inch I.D. tubing, size D hose connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty