Would you like to log in again?
Beakers, Pilot Plant, Ace Glass Incorporated
Supplier: Ace Glass
6228-05 6231-27 6228-10 6231-21
89054-040EA 723.8 USD InStock
89054-040 89054-042 89054-046 89054-048
Beakers, Pilot Plant, Ace Glass Incorporated
Beakers
This pilot plant beaker is used for large sizes for batch operations and mixing large volumes of measure liquids
  • Graduated
  • Fabricated from borosilicate glass
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty