Would you like to log in again?
Unjacketed Filter Reactor, Small Volume, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9443-10 9443-20 6381-30 6381-05 6381-10 6381-25
89074-564EA 561.64 USD InStock
89074-564 89054-080 89074-562 89054-082 89054-084 89054-086
Unjacketed Filter Reactor, Small Volume, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This small-volume filter reactor features a [ST] 45/50 joint at top, and a #50 Ace-Thred (internal glass thread) at bottom that accepts a size 50 PTFE bottom adapter (89052-600) with filter support and drain valve
Head has a [ST] 19/38 center neck and three [ST] 14/20 side necks.
Ordering information: Complete item consists of reactor body, head with [ST] joints, clamp, #50 PTFE bottom adapter with size –136 CAPFE O-Ring, 350µm polypropylene screen support, 100µm polyethylene support, adapter, shutoff valve (89052-616), non-flaking bearing (89061-866), stirring rod (89075-082), and blade (89075-100).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty