Would you like to log in again?
Peptide Vessel with Threaded Top, PTFE Stopcock Side Arm, and Removable Filter, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6407-32 6407-42 6407-48 6407-57 6407-08 6407-36 6407-54 6407-02 6407-15 6407-19 6407-12 6407-60 6407-11 6407-06
89054-228EA 639.2 USD InStock
89054-228 89054-216 89054-238 89054-218 89054-220 89054-230 89054-222 89054-232 89054-224 89054-234 89054-212 89054-226 89054-236 89054-214
Peptide Vessel with Threaded Top, PTFE Stopcock Side Arm, and Removable Filter, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical vessel is equipped with a 2mm bore PTFE stopcock, and a GL thread at top and Ace-Thred at bottom that accepts a PTFE adapter with removable filter disc and a PTFE shut-off valve
PTFE adapter has a "front-type" seal using a CAPFE (PTFE encapsulated silicone) O-Ring to make a compression seal in the Ace-Thred on vessel. Porosity C (25–50µm) glass filter disc is press fit into recess at top of the adapter and it can be replaced easily. Other end of adapter has a ¼" FNPT thread that allows connection of a 2.5mm bore PTFE shut-off valve with a luer extension. Supplied with a solid cap GL thread. Complete item consists of vessel, PTFE adapter, CAPFE O-Ring, and valve.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty