Would you like to log in again?
Peptide Vessel with Ace-Thred Top and Bottom, Ace Glass Incorporated
Supplier: Ace Glass
6408-06 6408-08 6408-04 6408-10 6408-02
89054-240EA 133.56 USD InStock
89054-240 89054-242 89054-244 89054-246 89054-248
Peptide Vessel with Ace-Thred Top and Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical vessels with #15 Ace-Thred at both ends
Vessels have no side port. Use PTFE adapter (89052-602 - not included) with removable filter disc and PTFE valve (89052-608 - not included) with Luer-Lok extension at either end.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty