Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pressure Reaction Flasks, Unjacketed, Ace Glass Incorporated
Supplier: Ace Glass
6423-07 6423-10 6423-20 6423-35 6423-30 6423-05
89054-282EA 1435.77 USD InStock
89054-282 89054-284 89054-250 89054-252 89054-254 89054-256
Pressure Reaction Flasks, Unjacketed, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
These rugged, heavy-wall, round-bottom flasks are for use with the two-piece Pressure Reactor (head is sold separately) and can be used under elevated pressures
Schott-type flange is supplied with a shallow O-ring groove for use with mating grooved head.
Ordering information: Supplied with CAPFE O-ring. For head, heating mantle, silicone O-rings, or other parts, contact your Avantor representative.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty