Would you like to log in again?
Two-Piece Jacketed Pressure Reactor, Ace Glass Incorporated
Supplier: Ace Glass
6423-205 6423-225 6423-210 6423-200 6423-220 6423-215
89054-260EA 16264.86 USD InStock
89054-260 89054-262 89054-264 89054-266 89054-268 89054-258
Two-Piece Jacketed Pressure Reactor, Ace Glass Incorporated
Laboratory Reactor Systems
The complete, two-piece, plain flask, pressure reactor system is designed to perform low to moderate positive pressure reactions, synthesis, and catalysis
Reactor capacities range from 500 mL to 5000 mL, and Ace-Threds are used to achieve a leak-tight system. The two-piece (flanged flask and head) design allows the use of large agitators and facilitates easy product removal and clean-out. Rated 36 psi at 100 °C.
Ordering information: Complete systems include PTFE plug, West condenser, flask, head, complete pressure reactor manifold, thermowell, clamp, addition funnel, PTFE bushings with O-Rings, bearing (complete with Ace-Threds), stirring shaft, flexible shaft, PTFE stirrer blade, Krytox high-vacuum grease, swivel coupling, aluminum heating mantle, type "J" PTFE-clad thermocouple sensor, digital temperature controller with "J" output, polypropylene tubing, solid state stirrer motor controller, solid state stirrer motor with mounting rod, and 1.27 cm (1/2") PTFE sealing tape.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty