Would you like to log in again?
Pressure Reactor, Two-Piece With Bottom Outlet, Ace Glass Incorporated
Supplier: Ace Glass
6425-221 6425-12 6425-15 6425-206 6425-04 6425-06 6425-226 6425-214 6425-201 6425-19 6425-211 6425-23
89054-304EA 15885.25 USD InStock
89054-304 89054-290 89054-306 89054-292 89054-308 89054-294 89054-286 89054-296 89054-288 89054-298 89054-300 89054-302
Pressure Reactor, Two-Piece With Bottom Outlet, Ace Glass Incorporated
Laboratory Reactor Systems
Two-Piece Pressure Reactor System for use at pressures up to 35 PSI at 100 Degrees C
Has threaded bottom outlet with ball joint bottom oultlet connection.
Ordering information: Complete Systems come Supplied With Valves and Ball Joint Bottom Outlet Glass Body, Schott Flange Head With #15 Center Neck,(2) #15 and (1) #7 Side Necks, Addition Funnel, Condenser, Manifold With Gauge and Rupture Disc, Stirring and Temperature Control Assemblies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty