Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Epoxy Coated Pressure Reactor Manifold, Ace Glass Incorporated
Supplier: Ace Glass
6448-68 6448-24 6448-54
89054-490EA 431.78 USD InStock
89054-490 89054-492 89054-494
Epoxy Coated Pressure Reactor Manifold, Ace Glass Incorporated
Manifolds
Pressure manifold provides for safer operation of Ace pressure filter reactors.
Manifold features an epoxy coating, 0-60 psi pressure gauge with stainless steel internals, 0-50 psi adjustable relief valve and over-pressure safety rupture disc. Ace-Thred to NPT connections are used throughout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty