Would you like to log in again?
Spherical Flask Reaction System, 50 L, Ace Glass Incorporated
Supplier: Ace Glass
6470-68
89054-542EA 27440.09 USD InStock
89054-542
Spherical Flask Reaction System, 50 L, Ace Glass Incorporated
Laboratory Reactor Systems
Comes with rugged 19 mm diameter, steel-reinforced PTFE stirring rod needs no lubrication
Spherical flask for a reaction assembly with a center neck and 2 side necks which are [SJ] 65/40, and two necks [ST] 29/42. Fabricated from borosilicate glass.
Thermowell without a joint with an outside diameter of 10 mm. The completely adjustable depth feature of this combination thermowell means the well is less expensive and more versatile than the jointed style.
Runs in a precision Trubore® bearing. The PTFE paddle is removable and other interchangeable agitators. Highly efficient condenser is attached directly to 65/40 side joint. Thermowell depth is completely adjustable via an Ace-Thred adapter. Unit as supplied has a heavy-duty mechanical lab stirrer; for situations that call for non-electric motors, ACE can supply a compact air-driven motor and support. Heavy-duty bottom heating mantle.
Ordering information: Complete Assembly includes 50 L flask, heating mantle and controls, mechanical stirrer and controls, pressure equalizing funnel and pilot plant condenser. All components can be purchased seperately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty