Would you like to log in again?
Unjacketed Cylindrical Reaction Flask, KF Plane Flange, Ace Glass Incorporated
Supplier: Ace Glass
6472-65 6472-64 6472-241 6472-248 6473-11 6473-05 6472-249 6472-242
89054-766EA 3333.72 USD InStock
89054-766 89054-656 89054-768 89054-658 89054-808 89054-818 89054-662 89054-664
Unjacketed Cylindrical Reaction Flask, KF Plane Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
These unjacketed cylindrical reaction flask are made of heavy wall borosilicate glass and include a bottom outlet valve
Reactor accessories including Flat Flange Couplings, Flat Flange PTFE Gaskets, Bottom Outlet Valves, Beaded Pipe Couplings for Valves, Spherical Joint Adapters for Valves, and Spherical Joint O-Rings are sold separately.
Each flask has high visibility capacity reference graduations for easy viewing. They can be used standalone or with bottom opening heating mantles. Available with Schott-Duran O-ring groove or KF Plane flat ground flanges. as well as a variety of drain valve styles to select from.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty